Assay Details
Gene Symbol: LRRC3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193981.2 | NP_001180910.1 | 207 | 101 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001193981.2 transcripts.
Set Membership: |
![]() ![]() |
Panther Classification:
