Assay Details
| Gene Symbol: GCNT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194134.2 | NP_001181063.2 | 1449 | 77 | ||
| XM_015142342.2 | XP_014997828.2 | 2451 | 77 | |||
| XM_015142345.2 | XP_014997831.2 | 2359 | 77 | |||
| XM_015142346.2 | XP_014997832.2 | 2128 | 77 | |||
More Information
Set Membership: |
|

GCNT3
Genomic Map