Assay Details
Gene Symbol: B3GALT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194204.1 | NP_001181133.1 | 1552 | 101 | ||
XM_015110249.2 | XP_014965735.1 | 1751 | 101 | |||
XM_015110250.2 | XP_014965736.1 | 2305 | 101 | |||
XM_028830504.1 | XP_028686337.1 | 1834 | 101 | |||
XM_028830505.1 | XP_028686338.1 | 1855 | 101 | |||
XM_028830506.1 | XP_028686339.1 | 1763 | 101 | |||
XM_028830507.1 | XP_028686340.1 | 1784 | 101 | |||
GenBank mRNA | JU328559.1 | - | 921 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
