Assay Details
| Gene Symbol: LRRC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194240.1 | NP_001181169.1 | 652 | 87 | ||
| XM_015132715.2 | XP_014988201.1 | 965 | 87 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LRRC3| Gene Symbol: LRRC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194240.1 | NP_001181169.1 | 652 | 87 | ||
| XM_015132715.2 | XP_014988201.1 | 965 | 87 | |||
Set Membership: |
|