Assay Details
| Gene Symbol: NDUFS6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194314.2 | NP_001181243.2 | 216 | 74 | ||
| GenBank mRNA | JV652326.1 | - | 189 | 74 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NDUFS6| Gene Symbol: NDUFS6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194314.2 | NP_001181243.2 | 216 | 74 | ||
| GenBank mRNA | JV652326.1 | - | 189 | 74 | ||
Set Membership: |
|