Assay Details
Gene Symbol: TSPAN10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194513.2 | NP_001181442.2 | 39 | 73 | ||
XM_015120545.2 | XP_014976031.2 | 459 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
