Assay Details
Gene Symbol: PAQR9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194535.3 | NP_001181464.2 | 2146 | 87 | ||
XM_028843359.1 | XP_028699192.1 | 2035 | 87 | |||
GenBank mRNA | JV713858.1 | - | 1770 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
