Assay Details
Gene Symbol: WDR5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194550.2 | NP_001181479.2 | 1249 | 77 | ||
GenBank mRNA | JU322738.1 | - | 1154 | 77 | ||
JU473555.1 | - | 1129 | 77 | |||
JV044765.1 | - | 1235 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
