Assay Details
Gene Symbol: SAPCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194586.1 | NP_001181515.1 | 173 | 99 | ||
XM_015135941.2 | XP_014991427.1 | 398 | 99 | |||
XM_015135943.2 | XP_014991429.1 | 398 | 99 | |||
XM_028846838.1 | XP_028702671.1 | 398 | 99 | |||
GenBank mRNA | JV651355.1 | - | 121 | 99 |
