Assay Details
Gene Symbol: SAPCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194586.1 | NP_001181515.1 | 486 | 64 | ||
XM_028846838.1 | XP_028702671.1 | 816 | 64 | |||
GenBank mRNA | JV651355.1 | - | 434 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
