Assay Details
Gene Symbol: TLCD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194730.2 | NP_001181659.2 | 259 | 82 | ||
XM_028835774.1 | XP_028691607.1 | 5747 | 82 | |||
XM_028835775.1 | XP_028691608.1 | 4344 | 82 | |||
XM_028835776.1 | XP_028691609.1 | 4259 | 82 | |||
XM_028835777.1 | XP_028691610.1 | 4967 | 82 | |||
XM_028835778.1 | XP_028691611.1 | 4820 | 82 | |||
XM_028835780.1 | XP_028691613.1 | 3425 | 82 | |||
XM_028835781.1 | XP_028691614.1 | 3328 | 82 |
