Assay Details
Gene Symbol: TMEM91 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194828.3 | NP_001181757.1 | 867 | 85 | ||
XM_015124016.2 | XP_014979502.1 | 635 | 85 | |||
XM_015124017.2 | XP_014979503.1 | 635 | 85 | |||
XM_015124018.2 | XP_014979504.1 | 635 | 85 | |||
XM_015124019.2 | XP_014979505.1 | 635 | 85 | |||
XM_015124020.2 | XP_014979506.1 | 635 | 85 | |||
XM_028838306.1 | XP_028694139.1 | 635 | 85 | |||
GenBank mRNA | JU321034.1 | - | 384 | 85 | ||
JV600313.1 | - | 557 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
