Assay Details
| Gene Symbol: TMEM170A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194844.1 | NP_001181773.1 | 414 | 65 | ||
| XM_015126635.2 | XP_014982121.1 | 265 | 65 | |||
| XM_015126636.2 | XP_014982122.1 | 483 | 65 | |||
| XM_028840327.1 | XP_028696160.1 | 203 | 65 | |||
| XM_028840328.1 | XP_028696161.1 | 352 | 65 | |||
| GenBank mRNA | JU327717.1 | - | 290 | 65 | ||
| JU474282.1 | - | 370 | 65 | |||
| JV635949.1 | - | 370 | 65 | |||
More Information
Set Membership: |
|

TMEM170A
Genomic Map