Assay Details
Gene Symbol: CLDN24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194880.2 | NP_001181809.1 | 53 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
