Assay Details
| Gene Symbol: APOC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195137.1 | NP_001182066.1 | 231 | 76 | ||
| GenBank mRNA | CO646055.1 | - | 233 | 76 | ||
| JU324955.1 | - | 134 | 76 | |||
More Information
Set Membership: |
|

APOC4
Genomic Map