Assay Details
Gene Symbol: MRPL57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193743.2 | NP_001180672.1 | 28 | 53 | ||
GenBank mRNA | JU332343.1 | - | 65 | 53 | ||
JU332344.1 | - | 65 | 53 | |||
JU471146.1 | - | 83 | 53 | |||
JV045425.1 | - | 83 | 53 | |||
JV529215.1 | - | 32 | 53 | |||
JV639260.1 | - | 87 | 53 | |||
JV704676.1 | - | 63 | 53 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001193743.2 transcripts.
Set Membership: |
![]() ![]() |
Panther Classification:
