Assay Details
Gene Symbol: C3H7orf50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193747.2 | NP_001180676.1 | 437 | 60 | ||
XM_015133126.2 | XP_014988612.1 | 542 | 60 | |||
GenBank mRNA | JU332125.1 | - | 461 | 60 | ||
JU472349.1 | - | 426 | 60 | |||
JV043321.1 | - | 418 | 60 | |||
JV638514.1 | - | 465 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
