Assay Details
Gene Symbol: NDUFB10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204758.1 | NP_001191687.1 | 330 | 97 | ||
GenBank mRNA | EG705474.1 | - | 330 | 97 | ||
JU334643.1 | - | 387 | 97 | |||
JU475703.1 | - | 319 | 97 | |||
JV047149.1 | - | 319 | 97 | |||
JV617613.1 | - | 398 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
