Assay Details
| Gene Symbol: TOX4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172413.1 | NP_001165884.1 | 926 | 101 | ||
| GenBank mRNA | JU320121.1 | - | 957 | 101 | ||
| JU478065.1 | - | 960 | 101 | |||
| JU478066.1 | - | 960 | 101 | |||
| JV599882.1 | - | 979 | 101 | |||
More Information
Set Membership: |
|

TOX4
Genomic Map