Assay Details
Gene Symbol: NR0B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195285.1 | NP_001182214.1 | 1173 | 110 | ||
GenBank mRNA | AF322896.1 | - | 1163 | 110 | ||
JV644414.1 | - | 343 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
