Assay Details
Gene Symbol: SFTPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195376.2 | NP_001182305.2 | 343 | 81 | ||
XM_015147128.2 | XP_015002614.2 | 270 | 81 | |||
XM_015147129.2 | XP_015002615.2 | 300 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
