Assay Details
Gene Symbol: ICAM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195749.1 | NP_001182678.1 | 341 | 101 | ||
XM_028838471.1 | XP_028694304.1 | 499 | 101 | |||
GenBank mRNA | AF340063.1 | - | 304 | 101 | ||
JU473373.1 | - | 388 | 101 |

More Information
Set Membership: |
![]() ![]() |
