Assay Details
| Gene Symbol: C6H16orf92 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194371.2 | NP_001181300.1 | 64 | 111 | ||
| XM_028840515.1 | XP_028696348.1 | 73 | 111 | |||
More Information
Set Membership: |
|

C6H16orf92
Genomic Map