Assay Details
Gene Symbol: ADM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194708.2 | NP_001181637.1 | 154 | 86 | ||
XM_002798424.3 | XP_002798470.1 | 489 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
