Assay Details
| Gene Symbol: TUBB4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194791.3 | NP_001181720.1 | 116 | 85 | ||
| XM_015116205.2 | XP_014971691.1 | 375 | 85 | |||
| GenBank mRNA | JV640111.1 | - | 140 | 85 | ||
More Information
Set Membership: |
|

TUBB4B
Genomic Map