Assay Details
Gene Symbol: PSENEN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194826.1 | NP_001181755.1 | 325 | 109 | ||
XM_002801186.3 | XP_002801232.1 | 397 | 109 | |||
XM_015123558.2 | XP_014979044.1 | 397 | 109 | |||
XM_028838309.1 | XP_028694142.1 | 363 | 109 | |||
XM_028838310.1 | XP_028694143.1 | 383 | 109 | |||
XM_028838311.1 | XP_028694144.1 | 345 | 109 | |||
XM_028838312.1 | XP_028694145.1 | 314 | 109 | |||
XM_028838313.1 | XP_028694146.1 | 331 | 109 | |||
GenBank mRNA | JU320462.1 | - | 375 | 109 | ||
JU473821.1 | - | 380 | 109 | |||
JV045648.1 | - | 344 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
