Assay Details
Gene Symbol: SULT1A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194840.2 | NP_001181769.2 | 244 | 95 | ||
XM_015125892.2 | XP_014981378.2 | 512 | 95 | |||
XM_015125893.2 | XP_014981379.2 | 230 | 95 | |||
XM_028840331.1 | XP_028696164.1 | 528 | 95 | |||
XM_028840332.1 | XP_028696165.1 | 526 | 95 | |||
GenBank mRNA | JV721957.1 | - | 207 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
