Assay Details
| Gene Symbol: CMPK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193618.1 | NP_001180547.1 | 952 | 126 | ||
| XM_028843986.1 | XP_028699819.1 | 901 | 126 | |||
| GenBank mRNA | CN647695.1 | - | 184 | 126 | ||
| JU321279.1 | - | 1021 | 126 | |||
| JU472782.1 | - | 1033 | 126 | |||
| JV045528.1 | - | 979 | 126 | |||
More Information
Set Membership: |
|

CMPK1
Genomic Map