Assay Details
| Gene Symbol: MRPL57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193743.2 | NP_001180672.1 | 549 | 92 | ||
| GenBank mRNA | JU332343.1 | - | 585 | 92 | ||
| JU332344.1 | - | 585 | 92 | |||
| JU471146.1 | - | 603 | 92 | |||
| JV045425.1 | - | 604 | 92 | |||
| JV529215.1 | - | 553 | 92 | |||
| JV639260.1 | - | 607 | 92 | |||
| JV704676.1 | - | 584 | 92 | |||
More Information
Set Membership: |
|

MRPL57
Genomic Map