Assay Details
| Gene Symbol: H2BFWT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193918.1 | NP_001180847.1 | 46 | 60 | ||
| XM_015127878.2 | XP_014983364.1 | 64 | 60 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

H2BFWT| Gene Symbol: H2BFWT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193918.1 | NP_001180847.1 | 46 | 60 | ||
| XM_015127878.2 | XP_014983364.1 | 64 | 60 | |||
Set Membership: |
|