Assay Details
| Gene Symbol: PAX9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040417.2 | NP_001035507.2 | 848 | 75 | ||
| XM_028850138.1 | XP_028705971.1 | 1323 | 75 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PAX9| Gene Symbol: PAX9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040417.2 | NP_001035507.2 | 848 | 75 | ||
| XM_028850138.1 | XP_028705971.1 | 1323 | 75 | |||
Set Membership: |
|