Assay Details
| Gene Symbol: APOBEC3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246231.1 | NP_001233160.1 | 189 | 120 | ||
| GenBank mRNA | JF714484.1 | - | 189 | 120 | ||
| JF831054.1 | - | 189 | 120 | |||
More Information
Set Membership: |
|

APOBEC3A
Genomic Map