Assay Details
| Gene Symbol: SNRPG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193440.2 | NP_001180369.1 | 421 | 166 | ||
| XM_015112526.2 | XP_014968012.1 | 564 | 166 | |||
| XM_028831473.1 | XP_028687306.1 | 324 | 166 | |||
| XM_028831474.1 | XP_028687307.1 | 427 | 166 | |||
| GenBank mRNA | JU332370.1 | - | 455 | 166 | ||
| JV045818.1 | - | 463 | 166 | |||
More Information
Set Membership: |
|

SNRPG
Genomic Map