Assay Details
Gene Symbol: TREX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002802809.3 | XP_002802855.1 | 1402 | 98 | ||
GenBank mRNA | JU325314.1 | - | 811 | 98 | ||
JU479571.1 | - | 812 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
