Assay Details
Gene Symbol: C2H3orf38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190960.1 | NP_001177889.1 | 134 | 97 | ||
XM_015128531.2 | XP_014984017.1 | 484 | 97 | |||
GenBank mRNA | JU342211.1 | - | 258 | 97 | ||
JU484908.1 | - | 447 | 97 | |||
JV648988.1 | - | 314 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
