Assay Details
Gene Symbol: C2H3orf38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190960.1 | NP_001177889.1 | 370 | 93 | ||
GenBank mRNA | JU342211.1 | - | 494 | 93 | ||
JU484908.1 | - | 683 | 93 | |||
JV648988.1 | - | 550 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() |
