Assay Details
Gene Symbol: CHST11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258082.1 | NP_001245011.1 | 607 | 80 | ||
XM_028828846.1 | XP_028684679.1 | 104 | 80 | |||
GenBank mRNA | JU475779.1 | - | 607 | 80 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028828846.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
