Assay Details
Gene Symbol: AURKAIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260532.1 | NP_001247461.1 | 562 | 53 | ||
XM_028843133.1 | XP_028698966.1 | 1753 | 53 | |||
XM_028843136.1 | XP_028698969.1 | 1672 | 53 | |||
XM_028843140.1 | XP_028698973.1 | 1514 | 53 | |||
GenBank mRNA | JU335182.1 | - | 597 | 53 | ||
JV045515.1 | - | 562 | 53 | |||
JV654847.1 | - | 607 | 53 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
