Assay Details
Gene Symbol: BOLA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257561.1 | NP_001244490.1 | 321 | 106 | ||
XM_015150500.2 | XP_015005986.1 | 371 | 106 | |||
XM_028842221.1 | XP_028698054.1 | 561 | 106 | |||
GenBank mRNA | JU332330.1 | - | 321 | 106 | ||
JU473818.1 | - | 380 | 106 | |||
JV045581.1 | - | 309 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
