Assay Details
Gene Symbol: TXNL4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257626.1 | NP_001244555.1 | 437 | 86 | ||
XM_015126569.2 | XP_014982055.2 | 466 | 86 | |||
XM_015126574.2 | XP_014982060.1 | 596 | 86 | |||
XM_028840484.1 | XP_028696317.1 | 644 | 86 | |||
XM_028840485.1 | XP_028696318.1 | 493 | 86 | |||
XM_028840486.1 | XP_028696319.1 | 268 | 86 | |||
XM_028840487.1 | XP_028696320.1 | 455 | 86 | |||
XM_028840488.1 | XP_028696321.1 | 466 | 86 | |||
XM_028840490.1 | XP_028696323.1 | 466 | 86 | |||
XM_028840491.1 | XP_028696324.1 | 466 | 86 | |||
XM_028840492.1 | XP_028696325.1 | 455 | 86 | |||
GenBank mRNA | JU322032.1 | - | 437 | 86 | ||
JU472948.1 | - | 442 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
