Assay Details
Gene Symbol: GEMIN8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257640.1 | NP_001244569.1 | 718 | 61 | ||
XM_015126975.2 | XP_014982461.1 | 875 | 61 | |||
XM_015126977.2 | XP_014982463.1 | 879 | 61 | |||
XM_015126978.2 | XP_014982464.1 | 705 | 61 | |||
GenBank mRNA | JU335169.1 | - | 718 | 61 | ||
JU472370.1 | - | 609 | 61 | |||
JV045302.1 | - | 724 | 61 | |||
JV703252.1 | - | 696 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
