Assay Details
| Gene Symbol: ETFRF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257657.1 | NP_001244586.1 | 120 | 105 | ||
| XM_015151132.2 | XP_015006618.1 | 159 | 105 | |||
| GenBank mRNA | JU320277.1 | - | 120 | 105 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001257657.1, XM_015151132.2 transcripts.
Set Membership: |
|

ETFRF1
Genomic Map