Assay Details
| Gene Symbol: GGPS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257690.1 | NP_001244619.1 | 302 | 113 | ||
| XM_015126452.2 | XP_014981938.1 | 380 | 113 | |||
| GenBank mRNA | JU337183.1 | - | 302 | 113 | ||
| JU477914.1 | - | 236 | 113 | |||
| JV048755.1 | - | 101 | 113 | |||
| JV598865.1 | - | 324 | 113 | |||
| JV666054.1 | - | 318 | 113 | |||
More Information
Set Membership: |
|

GGPS1
Genomic Map