Assay Details
Gene Symbol: SPSB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257717.1 | NP_001244646.1 | 905 | 65 | ||
GenBank mRNA | JU319836.1 | - | 905 | 65 | ||
JU471670.1 | - | 868 | 65 | |||
JV048301.1 | - | 863 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
