Assay Details
| Gene Symbol: LDHA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257735.2 | NP_001244664.1 | 330 | 78 | ||
| GenBank mRNA | JU321488.1 | - | 331 | 78 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LDHA| Gene Symbol: LDHA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257735.2 | NP_001244664.1 | 330 | 78 | ||
| GenBank mRNA | JU321488.1 | - | 331 | 78 | ||
Set Membership: |
|