Assay Details
| Gene Symbol: KCNIP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257864.1 | NP_001244793.1 | 295 | 83 | ||
| XM_015112830.2 | XP_014968316.1 | 402 | 83 | |||
| GenBank mRNA | JU322491.1 | - | 295 | 83 | ||
| JU332335.1 | - | 295 | 83 | |||
| JU472856.1 | - | 292 | 83 | |||
| JV602086.1 | - | 296 | 83 | |||
More Information
Set Membership: |
|

KCNIP3
Genomic Map