Assay Details
Gene Symbol: TCF21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260958.1 | NP_001247887.1 | 509 | 81 | ||
GenBank mRNA | JV045441.1 | - | 509 | 81 | ||
JV635440.1 | - | 495 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
