Assay Details
Gene Symbol: B3GALT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260985.1 | NP_001247914.1 | 915 | 108 | ||
GenBank mRNA | JU477635.1 | - | 915 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
