Assay Details
| Gene Symbol: CDKN1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261005.1 | NP_001247934.1 | 1077 | 92 | ||
| XM_028828821.1 | XP_028684654.1 | 1345 | 92 | |||
| GenBank mRNA | JU321114.1 | - | 1035 | 92 | ||
| JV044784.1 | - | 1077 | 92 | |||
| JV529662.1 | - | 1087 | 92 | |||
More Information
Set Membership: |
|

CDKN1B
Genomic Map