Assay Details
| Gene Symbol: RAD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261160.1 | NP_001248089.1 | 494 | 120 | ||
| XM_015139804.2 | XP_014995290.1 | 554 | 120 | |||
| XM_028849130.1 | XP_028704963.1 | 415 | 120 | |||
| XM_028849131.1 | XP_028704964.1 | 576 | 120 | |||
| XM_028849132.1 | XP_028704965.1 | 558 | 120 | |||
| GenBank mRNA | JU335711.1 | - | 482 | 120 | ||
| JU476596.1 | - | 494 | 120 | |||
More Information
Set Membership: |
|

RAD1
Genomic Map